Assay Details
| Gene Symbol: HSD11B1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001201.1 | NP_001001201.1 | 6 - 7 | 633 | 67 | |
| XM_015299692.1 | XP_015155178.1 | 5 - 6 | 891 | 67 | ||
| GenBank mRNA | AY578179.1 | - | 6 - 7 | 633 | 67 | |
More Information
Set Membership: |
|

HSD11B1L
Genomic Map