Assay Details
| Gene Symbol: IGFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001294.1 | NP_001001294.1 | 3 - 4 | 800 | 77 | |
| GenBank mRNA | AY591922.1 | - | 3 - 4 | 800 | 77 | |
| CR407577.1 | - | 3 - 4 | 733 | 77 | ||
More Information
Set Membership: |
|

IGFBP1
Genomic Map