Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001298.1 | NP_001001298.1 | 4 - 5 | 476 | 55 | |
| GenBank mRNA | AY615522.1 | - | 4 - 5 | 476 | 55 | |
| BX950581.1 | - | 4 - 5 | 486 | 55 | ||
More Information
Set Membership: |
|

PAH
Genomic Map