Assay Details
| Gene Symbol: TPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001301.1 | NP_001001301.1 | 7 - 8 | 973 | 63 | |
| XM_025147087.1 | XP_025002855.1 | 7 - 8 | 972 | 63 | ||
| XM_025147089.1 | XP_025002857.1 | 7 - 8 | 972 | 63 | ||
| XM_025147091.1 | XP_025002859.1 | 7 - 8 | 972 | 63 | ||
| GenBank mRNA | AY615523.1 | - | 7 - 8 | 973 | 63 | |
More Information
Set Membership: |
|

TPH2
Genomic Map