Assay Details
| Gene Symbol: KRT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001312.2 | NP_001001312.2 | 1 - 2 | 587 | 90 | |
| GenBank mRNA | AY574986.1 | - | 2 - 3 | 489 | 90 | |
| DR427178.1 | - | 1 - 2 | 587 | 90 | ||
More Information
Set Membership: |
|

KRT13
Genomic Map