Assay Details
| Gene Symbol: KRT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001313.2 | NP_001001313.2 | 2 - 3 | 767 | 131 | |
| GenBank mRNA | AB734680.1 | - | 2 - 3 | 678 | 131 | |
| AB734681.1 | - | 2 - 3 | 678 | 131 | ||
| AB734682.1 | - | 2 - 3 | 678 | 131 | ||
| AB734683.1 | - | 2 - 3 | 678 | 131 | ||
| AY574985.1 | - | 2 - 3 | 764 | 128 | ||
| JQ013796.1 | - | 2 - 3 | 678 | 131 | ||
More Information
Set Membership: |
|

KRT6A
Genomic Map