Assay Details
| Gene Symbol: KRT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001313.2 | NP_001001313.2 | 8 - 9 | 1466 | 133 | |
| GenBank mRNA | AB734681.1 | - | 8 - 9 | 1377 | 133 | |
| AB734682.1 | - | 8 - 9 | 1377 | 133 | ||
| AB734683.1 | - | 8 - 9 | 1377 | 133 | ||
| AY574985.1 | - | 8 - 9 | 1463 | 133 | ||
More Information
Set Membership: |
|

KRT6A
Genomic Map