Assay Details
| Gene Symbol: TEAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001337.1 | NP_001001337.1 | 8 - 9 | 892 | 75 | |
| NM_001001340.2 | NP_001001340.1 | 6 - 7 | 562 | 75 | ||
| NM_001001341.1 | NP_001001341.1 | 6 - 7 | 679 | 75 | ||
| NM_001306152.2 | NP_001293081.1 | 8 - 9 | 853 | 75 | ||
| NM_001359859.1 | NP_001346788.1 | 7 - 8 | 814 | 75 | ||
| NM_204771.1 | NP_990102.1 | 7 - 8 | 814 | 75 | ||
| GenBank mRNA | BX932327.1 | - | 6 - 7 | 679 | 75 | |
| U04834.1 | - | 7 - 8 | 777 | 75 | ||
| U06848.1 | - | 8 - 9 | 855 | 75 | ||
| U06849.1 | - | 8 - 9 | 816 | 75 | ||
| U06851.1 | - | 6 - 7 | 526 | 75 | ||
More Information
Set Membership: |
|

TEAD4
Genomic Map