Assay Details
| Gene Symbol: SDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001462.1 | NP_001001462.1 | 4 - 5 | 711 | 109 | |
| XM_025146997.1 | XP_025002765.1 | 5 - 6 | 576 | 109 | ||
| GenBank mRNA | AB105811.1 | - | 4 - 5 | 462 | 109 | |
| AF508228.1 | - | 4 - 5 | 711 | 109 | ||
| AF508229.1 | - | 4 - 5 | 711 | 109 | ||
More Information
Set Membership: |
|

SDC2
Genomic Map