Assay Details
| Gene Symbol: AMY2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001473.2 | NP_001001473.2 | 1 - 2 | 179 | 112 | |
| GenBank mRNA | CR406458.1 | - | 1 - 2 | 176 | 112 | |
More Information
Set Membership: |
|

AMY2A
Genomic Map