Assay Details
| Gene Symbol: AvBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001608.2 | NP_001001608.2 | 2 - 3 | 420 | 79 | |
| GenBank mRNA | AY621307.1 | - | 2 - 3 | 420 | 79 | |
More Information
Set Membership: |
|

AvBD5
Genomic Map