Assay Details
| Gene Symbol: AvBD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001609.2 | NP_001001609.1 | 1 - 2 | 131 | 67 | |
| GenBank mRNA | AY534896.1 | - | 1 - 2 | 55 | 67 | |
| AY621312.2 | - | 1 - 2 | 115 | 67 | ||
| BX274867.4 | - | 1 - 2 | 97 | 67 | ||
| CR388516.1 | - | 1 - 2 | 103 | 67 | ||
| DQ677641.1 | - | 1 - 2 | 81 | 67 | ||
| DQ858307.1 | - | 1 - 2 | 81 | 67 | ||
| DQ858320.1 | - | 1 - 2 | 81 | 67 | ||
| DQ858333.1 | - | 1 - 2 | 81 | 67 | ||
| DQ858347.1 | - | 1 - 2 | 81 | 67 | ||
More Information
Set Membership: |
|

AvBD10
Genomic Map