Assay Details
| Gene Symbol: AvBD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001611.2 | NP_001001611.1 | 1 - 2 | 220 | 73 | |
| GenBank mRNA | AY534894.1 | - | 1 - 2 | 58 | 73 | |
| AY621311.1 | - | 1 - 2 | 220 | 73 | ||
| DQ677640.1 | - | 1 - 2 | 88 | 73 | ||
| DQ858306.1 | - | 1 - 2 | 88 | 73 | ||
| DQ858319.1 | - | 1 - 2 | 88 | 73 | ||
| DQ858332.1 | - | 1 - 2 | 88 | 73 | ||
| KR136304.1 | - | 1 - 2 | 58 | 73 | ||
More Information
Set Membership: |
|

AvBD9
Genomic Map