Assay Details
Gene Symbol: ST8SIA3Z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001746.1 | NP_001001746.1 | 2 - 3 | 294 | 100 | |
GenBank mRNA | AJ699420.1 | - | 2 - 3 | 294 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
