Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001751.2 | NP_001001751.1 | 3 - 4 | 264 | 101 | |
| GenBank mRNA | AJ250337.1 | - | 3 - 4 | 265 | 101 | |
More Information
Set Membership: |
|

CYP3A5
Genomic Map