Assay Details
| Gene Symbol: CDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001760.1 | NP_001001760.1 | 10 - 11 | 1574 | 68 | |
| XM_015292521.2 | XP_015148007.1 | 10 - 11 | 1553 | 68 | ||
| XM_015292522.2 | XP_015148008.1 | 10 - 11 | 1649 | 68 | ||
| XM_015292523.2 | XP_015148009.1 | 10 - 11 | 1553 | 68 | ||
| XM_015292524.2 | XP_015148010.1 | 10 - 11 | 1631 | 68 | ||
| GenBank mRNA | M81779.1 | - | 10 - 11 | 1579 | 68 | |
| S62757.1 | - | 10 - 11 | 1574 | 68 | ||
More Information
Set Membership: |
|

CDH13
Genomic Map