Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001761.3 | NP_001001761.2 | 5 - 6 | 876 | 135 | |
| NM_001364699.1 | NP_001351628.1 | 6 - 7 | 848 | 135 | ||
| GenBank mRNA | J04047.1 | - | 5 - 6 | 840 | 135 | |
More Information
Set Membership: |
|

CYP19A1
Genomic Map