Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001879.1 | NP_001001879.1 | 6 - 7 | 869 | 112 | |
| GenBank mRNA | BX932720.1 | - | 6 - 7 | 874 | 112 | |
| CR353824.1 | - | 6 - 7 | 869 | 112 | ||
More Information
Set Membership: |
|

CYP4V2
Genomic Map