Assay Details
| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002242.1 | NP_001002242.1 | 11 - 12 | 1106 | 60 | |
| XM_015296419.2 | XP_015151905.1 | 11 - 12 | 1161 | 60 | ||
| XM_025142266.1 | XP_024998034.1 | 12 - 13 | 1543 | 60 | ||
| XM_025142267.1 | XP_024998035.1 | 11 - 12 | 1099 | 60 | ||
| XM_025142268.1 | XP_024998036.1 | 11 - 12 | 1098 | 60 | ||
| XM_025142269.1 | XP_024998037.1 | 11 - 12 | 1098 | 60 | ||
| XM_025142270.1 | XP_024998038.1 | 11 - 12 | 1099 | 60 | ||
| GenBank mRNA | M59456.1 | - | 11 - 12 | 1106 | 60 | |
More Information
Set Membership: |
|

CSNK2A1
Genomic Map