Assay Details
| Gene Symbol: IFI27L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002856.1 | NP_001002856.1 | 2 - 3 | 270 | 71 | |
| XM_015297642.2 | XP_015153128.1 | 3 - 4 | 347 | 71 | ||
| XM_015297643.2 | XP_015153129.1 | 3 - 4 | 411 | 71 | ||
| XM_015297644.2 | XP_015153130.1 | 4 - 5 | 3488 | 71 | ||
| XM_025142976.1 | XP_024998744.1 | 3 - 4 | 369 | 71 | ||
| GenBank mRNA | BX275934.4 | - | 2 - 3 | 264 | 71 | |
More Information
Set Membership: |
|

IFI27L2
Genomic Map