Assay Details
| Gene Symbol: RHCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004370.1 | NP_001004370.1 | 7 - 8 | 1187 | 65 | |
| GenBank mRNA | AY227357.1 | - | 7 - 8 | 1187 | 65 | |
More Information
Set Membership: |
|

RHCG
Genomic Map