Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004378.2 | NP_001004378.1 | 3 - 4 | 525 | 135 | |
| GenBank mRNA | AY393848.1 | - | 3 - 4 | 423 | 135 | |
| BU137661.1 | - | 2 - 3 | 268 | 135 | ||
| CB017418.1 | - | 2 - 3 | 166 | 135 | ||
| CR386189.1 | - | 3 - 4 | 505 | 135 | ||
| M24193.1 | - | 3 - 4 | 505 | 135 | ||
More Information
Set Membership: |
|

RACK1
Genomic Map