Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004378.2 | NP_001004378.1 | 5 - 6 | 738 | 61 | |
| GenBank mRNA | AY393848.1 | - | 5 - 6 | 636 | 61 | |
| BU137661.1 | - | 4 - 5 | 481 | 61 | ||
| CB017418.1 | - | 4 - 5 | 379 | 61 | ||
| CR386189.1 | - | 5 - 6 | 718 | 61 | ||
| M24193.1 | - | 5 - 6 | 718 | 61 | ||
More Information
Set Membership: |
|

RACK1
Genomic Map