Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004378.2 | NP_001004378.1 | 6 - 7 | 880 | 65 | |
| GenBank mRNA | AY393848.1 | - | 6 - 7 | 778 | 65 | |
| CK608329.1 | - | 2 - 3 | 263 | 65 | ||
| CR386189.1 | - | 6 - 7 | 860 | 65 | ||
| M24193.1 | - | 6 - 7 | 860 | 65 | ||
More Information
Set Membership: |
|

RACK1
Genomic Map