Assay Details
| Gene Symbol: RPL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004379.1 | NP_001004379.1 | 1 - 2 | 121 | 78 | |
| GenBank mRNA | CR385365.1 | - | 1 - 2 | 126 | 78 | |
| X62640.1 | - | 121 | 78 | |||
More Information
Set Membership: |
|

RPL7A
Genomic Map