Assay Details
| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004380.1 | NP_001004380.1 | 7 - 8 | 593 | 69 | |
| GenBank mRNA | M80589.1 | - | 7 - 8 | 593 | 69 | |
More Information
Set Membership: |
|

CAPZA2
Genomic Map