Assay Details
| Gene Symbol: IGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004384.2 | NP_001004384.1 | 2 - 3 | 469 | 70 | |
| GenBank mRNA | BU454475.1 | - | 2 - 3 | 387 | 70 | |
| FJ977570.1 | - | 2 - 3 | 411 | 70 | ||
| JN942578.1 | - | 1 - 2 | 61 | 70 | ||
| KC242239.1 | - | 2 - 3 | 223 | 70 | ||
| M32791.1 | - | 2 - 3 | 469 | 70 | ||
More Information
Set Membership: |
|

IGF1
Genomic Map