Assay Details
| Gene Symbol: STMN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004393.1 | NP_001004393.1 | 2 - 3 | 166 | 90 | |
| XM_025142225.1 | XP_024997993.1 | 2 - 3 | 182 | 90 | ||
| GenBank mRNA | AF076982.1 | - | 2 - 3 | 166 | 90 | |
More Information
Set Membership: |
|

STMN3
Genomic Map