Assay Details
| Gene Symbol: PPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004401.1 | NP_001004401.1 | 2 - 3 | 313 | 66 | |
| GenBank mRNA | CR388538.1 | - | 2 - 3 | 532 | 66 | |
| M60069.1 | - | 2 - 3 | 313 | 66 | ||
More Information
Set Membership: |
|

PPAT
Genomic Map