Assay Details
| Gene Symbol: VTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004408.2 | NP_001004408.1 | 14 - 15 | 2053 | 111 | |
| GenBank mRNA | D89547.1 | - | 14 - 15 | 2054 | 111 | |
More Information
Set Membership: |
|

VTG1
Genomic Map