Assay Details
| Gene Symbol: FKBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005431.1 | NP_001005431.1 | 531 | 60 | ||
| XM_015298716.2 | XP_015154202.1 | 614 | 60 | |||
| XM_025143713.1 | XP_024999481.1 | 2160 | 60 | |||
| GenBank mRNA | AJ719983.1 | - | 614 | 60 | ||
| AY723747.1 | - | 531 | 60 | |||
| CR385703.1 | - | 214 | 60 | |||
More Information
Set Membership: |
|

FKBP5
Genomic Map