Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006415.1 | NP_001006415.1 | 3 - 4 | 630 | 77 | |
| XM_025148781.1 | XP_025004549.1 | 3 - 4 | 667 | 77 | ||
| GenBank mRNA | AJ719453.1 | - | 3 - 4 | 630 | 77 | |
More Information
Set Membership: |
|

YWHAQ
Genomic Map