Assay Details
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204327.2 | NP_989658.1 | 5 - 6 | 832 | 150 | |
XM_015274146.2 | XP_015129632.1 | 6 - 7 | 877 | 150 | ||
GenBank mRNA | AF506010.2 | - | 5 - 6 | 830 | 150 | |
AJ721035.1 | - | 5 - 6 | 832 | 150 | ||
AY263378.1 | - | 5 - 6 | 752 | 150 | ||
EF122149.1 | - | 5 - 6 | 752 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
