Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204401.2 | NP_989732.2 | 3 - 4 | 782 | 127 | |
| GenBank mRNA | AJ720823.1 | - | 3 - 4 | 782 | 127 | |
| AY247742.1 | - | 3 - 4 | 782 | 127 | ||
More Information
Set Membership: |
|

TRIB2
Genomic Map