Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204427.2 | NP_989758.1 | 4 - 5 | 607 | 71 | |
| GenBank mRNA | AJ535754.1 | - | 4 - 5 | 607 | 71 | |
| AJ719585.1 | - | 4 - 5 | 565 | 71 | ||
| AJ720352.1 | - | 4 - 5 | 607 | 71 | ||
More Information
Set Membership: |
|

POFUT1
Genomic Map