Assay Details
| Gene Symbol: IL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204524.1 | NP_989855.1 | 5 - 6 | 623 | 92 | |
| XM_015297469.1 | XP_015152955.1 | 4 - 5 | 834 | 92 | ||
| GenBank mRNA | DQ393267.1 | - | 5 - 6 | 591 | 92 | |
| FJ788633.1 | - | 5 - 6 | 591 | 92 | ||
| HM179638.1 | - | 5 - 6 | 600 | 92 | ||
| HQ008778.1 | - | 5 - 6 | 591 | 92 | ||
| HQ329098.1 | - | 5 - 6 | 591 | 92 | ||
| Y15006.1 | - | 623 | 92 | |||
More Information
Set Membership: |
|

IL1B
Genomic Map