Assay Details
| Gene Symbol: IL2RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303432.1 | NP_001290361.1 | 1 - 2 | 200 | 102 | |
| NM_204527.2 | NP_989858.1 | 1 - 2 | 200 | 102 | ||
| GenBank mRNA | AJ419896.1 | - | 1 - 2 | 124 | 102 | |
| AJ419897.1 | - | 1 - 2 | 124 | 102 | ||
| CF074879.1 | - | 1 - 2 | 102 | |||
More Information
Set Membership: |
|

IL2RG
Genomic Map