Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204541.2 | NP_989872.1 | 3 - 4 | 380 | 68 | |
| GenBank mRNA | AB085851.1 | - | 3 - 4 | 339 | 68 | |
| AY584568.1 | - | 3 - 4 | 380 | 68 | ||
| BX934742.2 | - | 3 - 4 | 389 | 68 | ||
| EU049890.1 | - | 3 - 4 | 337 | 68 | ||
More Information
Set Membership: |
|

ORM1
Genomic Map