Assay Details
| Gene Symbol: ATVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204560.1 | NP_989891.1 | 7 - 8 | 1200 | 98 | |
| XM_015290126.2 | XP_015145612.1 | 7 - 8 | 1321 | 98 | ||
| XM_015290127.2 | XP_015145613.1 | 7 - 8 | 1152 | 98 | ||
| XM_015290128.2 | XP_015145614.1 | 7 - 8 | 1102 | 98 | ||
| XM_015290130.2 | XP_015145616.1 | 7 - 8 | 1160 | 98 | ||
| XM_015290131.2 | XP_015145617.1 | 7 - 8 | 1155 | 98 | ||
| XM_015290132.2 | XP_015145618.1 | 7 - 8 | 1184 | 98 | ||
| XM_015290133.2 | XP_015145619.1 | 7 - 8 | 1095 | 98 | ||
| XM_015290134.2 | XP_015145620.1 | 7 - 8 | 1149 | 98 | ||
| XM_025152083.1 | XP_025007851.1 | 7 - 8 | 1223 | 98 | ||
| XM_025152084.1 | XP_025007852.1 | 7 - 8 | 1099 | 98 | ||
| GenBank mRNA | AJ318064.2 | - | 7 - 8 | 1200 | 98 | |
| U38622.1 | - | 7 - 8 | 1162 | 98 | ||
More Information
Set Membership: |
|

ATVR1
Genomic Map