Assay Details
| Gene Symbol: CD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204649.1 | NP_989980.1 | 2 - 3 | 200 | 67 | |
| XM_015293164.2 | XP_015148650.1 | 2 - 3 | 237 | 67 | ||
| GenBank mRNA | AY528652.1 | - | 1 - 2 | 58 | 67 | |
| AY560013.1 | - | 1 - 2 | 58 | 67 | ||
| AY560014.1 | - | 1 - 2 | 58 | 67 | ||
| AY560016.1 | - | 1 - 2 | 58 | 67 | ||
| DQ202315.1 | - | 1 - 2 | 64 | 67 | ||
| Y12012.1 | - | 200 | 67 | |||
More Information
Set Membership: |
|

CD4
Genomic Map