Assay Details
| Gene Symbol: CD40LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204733.1 | NP_990064.1 | 2 - 3 | 367 | 79 | |
| GenBank mRNA | AJ243435.2 | - | 2 - 3 | 367 | 79 | |
| EF554722.1 | - | 2 - 3 | 328 | 79 | ||
More Information
Set Membership: |
|

CD40LG
Genomic Map