Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204817.1 | NP_990148.1 | 7 - 8 | 1196 | 98 | |
| XM_015284813.2 | XP_015140299.1 | 7 - 8 | 1347 | 98 | ||
| XM_015284814.2 | XP_015140300.1 | 6 - 7 | 1123 | 98 | ||
| GenBank mRNA | AJ131923.1 | - | 7 - 8 | 1196 | 98 | |
More Information
Set Membership: |
|

ANGPT2
Genomic Map