Assay Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204932.1 | NP_990263.1 | 6 - 7 | 1106 | 67 | |
| GenBank mRNA | AB734688.1 | - | 6 - 7 | 1049 | 67 | |
| AB734689.1 | - | 6 - 7 | 1049 | 67 | ||
| AF072698.1 | - | 6 - 7 | 1106 | 67 | ||
More Information
Set Membership: |
|

KRT7
Genomic Map