Assay Details
| Gene Symbol: ANGPT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204933.3 | NP_990264.3 | 2 - 3 | 159 | 61 | |
| XM_015295673.2 | XP_015151159.1 | 2 - 3 | 198 | 61 | ||
| GenBank mRNA | AF059262.1 | - | 2 - 3 | 61 | ||
More Information
Set Membership: |
|

ANGPT1L
Genomic Map