Assay Details
| Gene Symbol: LHCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204936.1 | NP_990267.1 | 7 - 8 | 656 | 68 | |
| GenBank mRNA | AB009283.1 | - | 7 - 8 | 656 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHCGR| Gene Symbol: LHCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204936.1 | NP_990267.1 | 7 - 8 | 656 | 68 | |
| GenBank mRNA | AB009283.1 | - | 7 - 8 | 656 | 68 | |
Set Membership: |
|