Assay Details
| Gene Symbol: COCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204937.1 | NP_990268.1 | 8 - 9 | 780 | 61 | |
| XM_015287284.2 | XP_015142770.1 | 8 - 9 | 925 | 61 | ||
| GenBank mRNA | AF012252.1 | - | 8 - 9 | 780 | 61 | |
More Information
Set Membership: |
|

COCH
Genomic Map