Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204940.2 | NP_990271.2 | 7 - 8 | 1105 | 84 | |
| XM_015284045.2 | XP_015139531.2 | 7 - 8 | 1084 | 84 | ||
| GenBank mRNA | U25176.1 | - | 7 - 8 | 1105 | 84 | |
| U67086.1 | - | 7 - 8 | 1037 | 84 | ||
More Information
Set Membership: |
|

TBXT
Genomic Map