Assay Details
| Gene Symbol: LFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204948.1 | NP_990279.1 | 2 - 3 | 502 | 57 | |
| GenBank mRNA | U91849.1 | - | 2 - 3 | 502 | 57 | |
| U97157.1 | - | 2 - 3 | 488 | 57 | ||
More Information
Set Membership: |
|

LFNG
Genomic Map