Assay Details
| Gene Symbol: TGFB1I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204984.1 | NP_990315.1 | 3 - 4 | 400 | 65 | |
| XM_015275206.2 | XP_015130692.1 | 3 - 4 | 899 | 65 | ||
| XM_015275207.2 | XP_015130693.1 | 3 - 4 | 899 | 65 | ||
| XM_015275208.2 | XP_015130694.1 | 3 - 4 | 899 | 65 | ||
| XM_015275209.2 | XP_015130695.1 | 3 - 4 | 520 | 65 | ||
| XM_015275210.2 | XP_015130696.1 | 3 - 4 | 366 | 65 | ||
| XM_015275211.2 | XP_015130697.1 | 3 - 4 | 367 | 65 | ||
| XM_015275213.1 | XP_015130699.1 | 3 - 4 | 899 | 65 | ||
| XM_015275215.2 | XP_015130701.1 | 3 - 4 | 821 | 65 | ||
| XM_015275216.2 | XP_015130702.1 | 3 - 4 | 899 | 65 | ||
| GenBank mRNA | L30099.1 | - | 3 - 4 | 400 | 65 | |
| U14589.1 | - | 3 - 4 | 383 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015275215.2 transcripts.
Set Membership: |
|

TGFB1I1
Genomic Map