Assay Details
| Gene Symbol: PGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204985.2 | NP_990316.1 | 2 - 3 | 217 | 102 | |
| GenBank mRNA | L37101.1 | - | 2 - 3 | 217 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGK2| Gene Symbol: PGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204985.2 | NP_990316.1 | 2 - 3 | 217 | 102 | |
| GenBank mRNA | L37101.1 | - | 2 - 3 | 217 | 102 | |
Set Membership: |
|